Learn More
Tolfenamic acid, 99+%, Thermo Scientific™
NSAID that activates calcium-activated potassium channels
82.65€ - 136.00€
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15463869
SDS Ver documentos
Thermo Scientific Alfa Aesar
J61256.06
ONLINE EXCLUSIVE PRICE
91.50€
82.65€ / 5 g
Ahorro 8.85€
9% Descuento
Ver productos alternativos
Ahorre hasta 
»
15473869
SDS Ver documentos
Thermo Scientific Alfa Aesar
J61256.09
10g
136.00€
10 g
Ver productos alternativos
Ahorre hasta 
»
Descripción
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Soluble in water (slightly), acetone (∼10 mg/ml), DMSO (52 mg/ml at 25°C), methanol (∼10 mg/ml) and ethanol (50 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.