Learn More
TOFA, MedChemExpress
Product Code. 16473248
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473248 10 mg 10mg
16449796 50 mg 50mg
16444516 100 mg 100mg
16435977 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473248 Supplier Medchem Express Supplier No. HY101068
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress TOFA (RMI14514;MDL14514) is an allosteric inhibitor of acetyl-CoA carboxylase-α (ACCA ).
TOFA (RMI14514
Specifications
Chemical Name or Material TOFA
Physical Form Solid
CAS 54857-86-2
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H32O4
For Use With (Application) Cancer-programmed cell death
Synonym RMI14514 MDL14514
Solubility Information DMSO : 11.36 mg/mL (35.01 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(OCCCCCCCCCCCCCC)O1)O
Molecular Weight (g/mol) 324.45
Formula Weight 324.45
Percent Purity 98.0%
Grade Research