Learn More
Unity Lab Services™ Thermo Scientific™ Ultra-Low Temperature Freezers Validation Documention
Product Code. 12704493
Change view
Click to view available options
For Use With (Equipment):
ThermoScientific ULT Freezers
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
12704493 ThermoScientific ULT Freezers Each
1 options
This item is not returnable. View return policy
Product Code. 12704493 Supplier Unity Lab Services™ Supplier No. IOQDOCE89003481
 Restricted Item
Fecha estimada de envoi: 21-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
OEM service provider. All protocols designed to optimize efficiency are included with each Ultra-Low Temperature Freezers Validation Documention for Forma 900 series, HeraFreeze HFUB and HFC , Thermo Scientific TSC and TSE, Revco ExF, Revco CxF, Thermo Scientific ULT10140 and ULT7150
  • IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ThermoScientific ULT Freezers
Description Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, open door and power loss testing.
Product Type IQ/OQ Documentation
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes IQ/OQ Documentation