Learn More
Unity Lab Services™ Thermo Scientific™ Centrifugation Validation Service for RDE, TDE, FDE and IUE Series
Product Code. 15815528
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Centrifugation
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15815528 Thermo Scientific Centrifugation Each
1 options
This item is not returnable. View return policy
Product Code. 15815528 Supplier Unity Lab Services™ Supplier No. IOQPCKE89003493
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Fecha estimada de envoi: 15-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Thermo Scientific Centrifugation Validation Service for Superspeed and Ultra centrifuges: Thermo Scientific, Sorvall
IQ
  • Unpacking, Assembly and Installation Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
  • Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ
  • System Component Information Detailed configuration information for each system component have been recorded.
  • OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
  • Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment.
Key Tests: Min/Max Speed, Brief/Extended Time, Temperature for Refrigerated Models
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Centrifugation
Product Type IQ/OQ Service
Service Offering Industry-Exclusive, No-Charge Requalification Guarantee
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service