Learn More
tert-Butyl-4-methoxyphenol, 99%, BLD Pharm™
Product Code. 30941432
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
10 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
100mg
10g
1kg
25g
500g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30941432 100 g 100g
30932491 25 g 25g
30938214 1 kg 1kg
30932495 500 g 500g
30937058 10 g 10g
30936611 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30941432 Supplier BLD Pharm Supplier No. BD119189100g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 25013-16-5
Molecular Formula C11H16O2
MDL Number MFCD01779059
Quantity 100 g
SMILES OC1=CC=C(C=C1C(C)(C)C)OC.OC2=CC=C(C(C(C)(C)C)=C2)OC
Molecular Weight (g/mol) 180.24
Percent Purity 99%
Chemical Name or Material tert-Butyl-4-methoxyphenol