Learn More
Dostmann Electronic™ Temperature Humidity Instrument
Product Code. 10543113
Change view
Click to view available options
Dimensions (L x W x H):
130 x 70 x 25 mm
170 x 50 x 17 mm
Unit Size:
Each
Product Code. Dimensions (L x W x H) unitSize
10543113 170 x 50 x 17 mm Each
10736840 130 x 70 x 25 mm Each
2 options
This item is not returnable. View return policy
Product Code. 10543113
Brand: Dostmann Electronic™ 50200560
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
This robust, reliable, ready-to-use temperature humidity instrument is a perfectly designed instrument for climatic measurements (HVAC branch).
  • Perfectly designed instrument for climatic meas­ure­ments (HVAC branch). 
    • The measuring instruments are perfect for measuring room climate (HVAC)
    • For detecting critical temperature-humidity environments: moul-risk in living quarters
    • The flexible combination sensor of P330 is excellent useable for measurements at hard-to-access points
Specifications
Type Thermohygrometer
Humidity Range 0 to 100%
Accuracy ±3%
Temperature Range (Metric) -40°C to +70°C
Temperature Accuracy ±0.5°C
Display Type LCD
Material Plastic
Dimensions (L x W x H) 170 x 50 x 17 mm
Weight (Metric) 130 g