Learn More
MedChemExpress SYBR Green qPCR Master Mix
Specifications
Specifications
| Content And Storage | Stored at -20°C protecting from light, and is stable for up to 18 months. Avoid repetitive freeze-thaw cycles while using. For immediate use, components may be stored at 2°C to 8°C. |
| Product Type | Kits |
| Quantity | 500 Reactions |
| Shipping Condition | Room temperature |