Learn More
STEMCELL Technologies STEMdiff™ Cardiomyocyte Freezing Medium
Product Code. 16337305
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Product Code. Quantity Unit Size
16337305 50 mL 50mL
1 options
This item is not returnable. View return policy
Product Code. 16337305 Supplier STEMCELL Technologies Supplier No. 05030
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Medium for cryopreserving hPSC-derived cardiomyocytes
  • STEMdiff™ Cardiomyocyte Freezing Medium can be used to cryopreserve cardiomyocytes derived from human pluripotent stem cells (hPSCs) and maintained in STEMdiff™ Cardiomyocyte Maintenance Medium (Catalog #05020)
  • STEMdiff™ Cardiomyocyte Dissociation Kit (Catalog #05025) is required for dissociating cardiomyocytes prior to freezing
  • After thawing using STEMdiff™ Cardiomyocyte Support Medium (Catalog #05027), the cardiomyocytes can be used in various downstream applications and analyses
  • This medium supports hPSC-derived cardiomyocytes generated using STEMdiff™ Cardiomyocyte Differentiation Kit (Catalog #05010)
Specifications
Cell Type Cardiomyocytes, PSC-Derived
For Use With (Application) Cryopreservation
Quantity 50 mL