Learn More
Hamilton™ Custom Kel-F Hub Needles
Product Code. 11532445
Change view
Click to view available options
Needle Gauge:
10 G
14 G
16 G
17 G
20 G
22 G
22s G
25 G
28 G
30 G
Needle Point Style:
2, 3, or 4
5 or AS
Custom
Unit Size:
Pack of 6
Product Code. Needle Gauge Needle Point Style unitSize
11532445 16 G 2, 3, or 4 Pack of 6
10516985 10 G 5 or AS Pack of 6
11562445 20 G 2, 3, or 4 Pack of 6
10004214 22 G 5 or AS Pack of 6
11532455 30 G 2, 3, or 4 Pack of 6
10779375 28 G 2, 3, or 4 Pack of 6
11590072 14 G Custom Pack of 6
11502455 25 G 2, 3, or 4 Pack of 6
11513274 22s G 5 or AS Pack of 6
11542445 17 G 2, 3, or 4 Pack of 6
10 options
This item is not returnable. View return policy
Product Code. 11532445
Brand: Hamilton™ 775007
This item is not returnable. View return policy
Design controlled by a series of universal standards which guarantees compatibility between manufacturers.
  • Available in various gauges
  • Provided in packs of 6
  • Custom point style
  • Autoclavable
  • Gas sterilizable
  • For Kel-F hubs
  • Custom length
  • Stainless steel Luer tip
Specifications
Autoclavable Not Autoclavable
Material Stainless Steel
Needle Gauge 16 G
Diameter (Metric) Inner 1.19 mm
Diameter (Metric) Outer 1.65 mm
Product Type Luer Lock Kel-F Hub Needle
Needle Point Style 2, 3, or 4
For Use With (Equipment) LT and TLL Syringes and Connectors
Connection Type Standard Tapered