Learn More
Alpaco™ Stainless Steel Clamps with Vertical Screws
Product Code. 11909677
Change view
Click to view available options
Size (Metric) Grip:
12 to 17 mm
14 to 19 mm
16 to 21 mm
17 to 22 mm
20 to 25 mm
22 to 27 mm
25 to 30 mm
28 to 33 mm
30 to 35 mm
36 to 41 mm
5 to 10 mm
7 to 12 mm
9 to 14 mm
Unit Size:
Pack of 10
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Grip unitSize
11909677 25 to 30 mm Pack of 10
11949657 5 to 10 mm Pack of 10
11979667 17 to 22 mm Pack of 10
11989667 20 to 25 mm Pack of 10
11999667 22 to 27 mm Pack of 10
11919677 28 to 33 mm Pack of 10
11919687 30 to 35 mm Pack of 10
11939687 36 to 41 mm Pack of 10
11949667 12 to 17 mm Pack of 10
11959667 14 to 19 mm Pack of 10
11969667 16 to 21 mm Pack of 10
11939717 7 to 12 mm Pack of 10
11949717 9 to 14 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 11909677 Supplier Alpaco™ Supplier No. INH 30/101631
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Provides uniform tightening around tubing for a watertight seal. Alpaco™ Stainless Steel Clamps with Vertical Screws are guaranteed waterproof and hand-lock without any tools.
  • Reusable
  • Resists pressure
  • No lubricant required for assembly
  • Smooth clamping to avoid tubing damage
Specifications
Quantity 10/Pk.
Type Hose Clamp
Material Stainless Steel
Size (Metric) Grip 25 to 30 mm