Description
- Rectangular, shallow stainless steel bowl with robust workmanship and high functionality
- Made of high-quality stainless steel 1.4301(304) with excellent corrosion resistance
- Ideal for demanding environments where hygiene is critical, such as laboratories, food processing facilities, and pharmaceutical plants
- Low design allows easy insertion and removal of instruments or samples and improves overview and accessibility
- Provides ample space for storing, sorting, or transporting utensils despite compact dimensions
- Smooth, easy-to-clean surface meets high hygiene standards and supports safe working practices
- Robust, durable, and corrosion-resistant construction
- Low, rectangular shape optimized for ergonomic working and quick access
- Smooth surface ensures easy cleaning and maximum hygiene
- Versatile in use
Specifications
Specifications
| Capacity (Metric) | 3000 mL |
| Dimensions (L x W x H) | 400 x 200 x 50 mm |
| Material | Stainless Steel |
| Type | Tray |
| Shape | Low Form, Rectangular |