missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate decahydrate, 99+%

Product Code. 11413247
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413247 10,000 g 10000g
11493237 500 g 500g
11403247 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413247 Supplier Thermo Scientific Alfa Aesar Supplier No. A16176.0C
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1303-96-4 | B4Na2O7·10H2O

It is used in detergents, cosmetics, and enamel glazes. It is likewise employed to prepare buffer solutions in biochemistry, as a fire retardant, anti-fungal compound, in the manufacture of fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a harbinger for other boron compounds. The molten borax bath is useful for carbide coating process on metals. It is applied as an efficient metal-free catalyst for Hetero-Michael reactions in an aqueous medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in detergents, cosmetics, and enamel glazes. It is likewise employed to prepare buffer solutions in biochemistry, as a fire retardant, anti-fungal compound, in the manufacture of fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a harbinger for other boron compounds. The molten borax bath is useful for carbide coating process on metals. It is applied as an efficient metal-free catalyst for Hetero-Michael reactions in an aqueous medium.

Solubility
Soluble in water, glycerol and ethanol. Insoluble in acids

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4Na2O7·10H2O
MDL Number MFCD00149193
Synonym Borax; Sodium borate (tetra) decahydrate

Specifications

Melting Point 75°C
pH 9.24
Quantity 10,000 g
Linear Formula Na2B4O7·10H2O
Merck Index 14,8590
Solubility Information Soluble in water,glycerol and ethanol. Insoluble in acids
Formula Weight 381.37 (201.22 Anhydrous)
Percent Purity ≥99%
Odor Odorless
Density 1.73
Chemical Name or Material Sodium tetraborate decahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.