Specifications
| Product Type | Cutting Mill |
| Certifications/Compliance | CE Certified |
| Description | Cutting Mill, 1675 H x 582 W x 700mm D, 220/230V, 50/60Hz, SM 100 Model, with Frequency Converter, 1, 500 min-1 Speed, 129.5mm dia. Rotor, 3-phase Power Connection, 1, 5 kW Drive Power, 3-phase Motor Drive, IP 54, <60 x 80mm Feed Size, Parallel Section Rotor/6-disc Rotor |
| Dimensions (L x W x H) | 700 x 582 x 1675 mm |
| Electrical Requirements | 220/230 V, 50/60 Hz |
| For Use With (Application) | For heavy-metal-free size reduction |
| Height (Metric) | 1675 mm |
| Hertz | 50/60 Hz |
| IP Rating | IP54 |
| Length (Metric) | 700 mm |
| Show More |