missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 98%

Product Code. 11313007
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313007 25 g 25g
11392997 1 g 1g
11303007 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11313007 Supplier Thermo Scientific Alfa Aesar Supplier No. L16322.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41851-59-6 | C9H13NO | 151.209 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41851-59-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00671660
InChI Key JTDGKQNNPKXKII-ZETCQYMHSA-N
Synonym s---1-4-methoxyphenyl ethylamine, s-1-4-methoxyphenyl ethanamine, s-1-4-methoxyphenyl ethylamine, s-1-4-methoxy-phenyl-ethylamine, 1s-1-4-methoxyphenyl ethanamine, s--1-4-methoxyphenyl ethylamine, s-p-1-4-methoxyphenyl ethylamine, s-1-4-methoxyphenyl ethan-1-amine, 1s-1-4-methoxyphenyl ethan-1-amine
PubChem CID 793467
IUPAC Name (1S)-1-(4-methoxyphenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)OC)N

Specifications

Melting Point <-20°C
Density 1.02
Boiling Point 65°C (0.38 mmHg)
Refractive Index 1.533
Quantity 25 g
UN Number UN2735
Beilstein 3196456
Sensitivity Air Sensitive
Optical Rotation −34° (Neat)
Formula Weight 151.21
Percent Purity ≥99%
Assay ee 98%
Chemical Name or Material (S)-(-)-1-(4-Methoxyphenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.