Learn More
Foxx Life Sciences Round PP Vacuum Trap System without Container
Product Code. 31933396
Encompass_Preferred
Change view
Click to view available options
Capacity (Metric):
2 L
4 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31933396 2 L Each
31933369 4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31933396 Supplier Foxx Life Sciences Supplier No. 3052004FLS
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Designed to meet strict CDC laboratory guidelines for handling biohazardous waste
  • Features:
    • Vacuum trap assembly with autoclavable PP bottles
    • 40 CFR 264.175 compliant aspirator system has a compact, tip-resistant secondary container for stability
    • Wide mouth PP bottles allow easy and thorough cleaning
    • Standard vacuum trap with 53B VersaCap™ and 83B VersaCap™ closure system with rotating adapter prevents tube tangling
    • Primary bottle volume: 2L, Overflow Bottle Volume: NA
  • Applications
    • Aspiration of cell culture media
    • Vacuum pump trap kit
    • Waste collection system for high-purity processes
    • Ideal for house vacuum or vacuum pump trap systems
    • Aspirator bottle assembly
Specifications
Capacity (Metric) 2 L
Material Polypropylene
Maximum Pressure 1 bar
Includes Bottle, Cap Assembly with Vent Filter, Tubing, Pinch Clamp
Type Round PP Vacuum Trap System