missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Heme-binding protein 1

Product Code. 16043691
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. Quantity unitSize
16043691 50 μg 50µg
16003691 10 μg 10µg
16093681 100 μg 100µg
16023691 200 μg 200µg
16033691 500 μg 500µg
16013691 1 mg 1mg
6 options
This item is not returnable. View return policy

Product Code. 16043691

Brand: enQuireBio™ QP8272ec50ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Heme-binding protein 2, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_055135.1
Gene ID (Entrez) 23593
Name Heme-binding protein 1
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol HEBP2
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence AEPLQPDPGA AEDAAAQAVE TPGWKAPEDA GPQPGSYEIR HYGPAKWVST SVESMDWDSA IQTGFTKLNS YIQGKNEKEM KIKMTAPVTS YVEPGSGPFS ESTITISLYI PSEQQFDPPR PLESDVFIED RAEMTVFVRS FDGFSSAQKN QEQLLTLASI LREDGKVFDE KVYYTAGYNS PVKLLNRNNE VWLIQKNEPT KENE
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.