Learn More
Radafaxine hydrochloride, MedChemExpress
Product Code. 16477408
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477408 10 mg 10mg
16439887 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16477408 Supplier Medchem Express Supplier No. HY17590
 Restricted Item
Fecha estimada de envoi: 24-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Radafaxine hydrochloride (GW-353162A) is a DAT (dopamine transporter) and NET(norepinephrine transporter) transporters inhibitor, and nAChR family modulator.
Radafaxine hydrochloride (GW-353162A) is a DAT (dopamine transporter) and NET(norepinephrine transporter) transporters inhibitor, and nAChR family modulator.
Specifications
Chemical Name or Material Radafaxine hydrochloride
Physical Form Solid
CAS 106083-71-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C13H19Cl2NO2
For Use With (Application) Neuroscience-Neuromodulation
Synonym GW-353162A BW-306U
Solubility Information DMSO : ≥ 38 mg/mL (130.05 mM) ∣H2O : 33.33 mg/mL (114.07 mM; Need ultrasonic)
SMILES O[C@]1(C2=CC=CC(Cl)=C2)[C@H](C)NC(C)(C)CO1.[H]Cl
Molecular Weight (g/mol) 292.2
Formula Weight 292.2
Percent Purity 99.88%
Grade Research