Description
- For use with freezers
- Includes locking rod
Specifications
Specifications
| For Use With (Equipment) | Freezers |
| Depth (Metric) | 140 mm |
| Includes | Locking rod |
| Shelves | 3 Tiers |
| Height (Metric) | 560 mm |
| Width (Metric) | 145 mm |