Learn More
Pyrene, 97%, BLD Pharm™
Product Code. 31091804
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31091804 10 g
31079055 1 kg
31085358 500 g
31090438 5 g
31078234 25 g
31079157 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31091804 Supplier BLD Pharm Supplier No. BD14965510g
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 129-00-0
Molecular Formula C16H10
MDL Number MFCD00004136
Quantity 10 g
SMILES C12=CC=C3C=CC=C4C=CC(C2=C34)=CC=C1
Molecular Weight (g/mol) 202.25
Percent Purity 97%
Chemical Name or Material Pyrene