Learn More
Phenoxyacetonitrile, 98%, BLD Pharm™
Product Code. 30985458
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30985458 5 g -
30972783 25 g 25g
30980184 100 g -
30969709 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30985458 Supplier BLD Pharm Supplier No. BD1475085g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 3598-14-9
Molecular Formula C8H7NO
MDL Number MFCD00052017
Quantity 5 g
SMILES N#CCOC1=CC=CC=C1
Molecular Weight (g/mol) 133.15
Percent Purity 98%
Chemical Name or Material Phenoxyacetonitrile