missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Peptone, from meat (pork gelatine)

Product Code. 15559463 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
Product Code. Quantity unitSize
15559463 5 kg 5kg
15569463 250 g 250g
15549463 1 kg 1kg
3 options
This item is not returnable. View return policy

Product Code. 15559463

Brand: Thermo Scientific Chemicals 458930050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Peptone, from meat (pork gelatine), CAS # 91079-38-8, is a nutrient isolated from meat used to promote the in vitro growth of several species including bacteria and fungi. | CAS: 91079-38-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

● Peptone, from meat, is a growth-promoting nutrient used in bacteria and fungi cultures

Applications

● It is commonly used in animal cell culture.

● Peptone is used in mass scale organism cultivation, and fermentation processes

● Peptone, from meat, is used as a supplement to in vitro culture media to help to increase cell number, specific monoclonal antibody production, and maintain long-term cultures

● This compound has been used as a component in Ax-2 axenic medium to induce reproduction of Dictyostelium discoideum

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91079-38-8

Specifications

Quantity 5 kg
Solubility Information Solubility aq. soln.: soluble.
Chemical Name or Material Peptone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.