Learn More
Pentafluorophenyl trifluoroacetate, 98%, BLD Pharm™
Especificaciones
Especificaciones
| CAS | 14533-84-7 |
| Molecular Formula | C8F8O2 |
| MDL Number | MFCD00134438 |
| Quantity | 25 g |
| SMILES | O=C(OC1=C(F)C(F)=C(F)C(F)=C1F)C(F)(F)F |
| Molecular Weight (g/mol) | 280.07 |
| Percent Purity | 98% |
| Chemical Name or Material | Pentafluorophenyl trifluoroacetate |