Learn More
PAβN dihydrochloride, MedChemExpress
Product Code. 16493368
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493368 10 mg 10mg
16489896 50 mg 50mg
16479845 250 mg 250mg
16462736 25 mg 25mg
16436097 5 mg 5mg
16414616 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16493368 Supplier Medchem Express Supplier No. HY101444A
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress PAβN dihydrochloride (MC-207110 dihydrochloride) is an efflux pump inhibitor.
PAβN dihydrochloride (MC-207110 dihydrochloride) is an efflux pump inhibitor.
Specifications
Chemical Name or Material PAβN dihydrochloride
Physical Form Solid
CAS 100929-99-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H32Cl2N6O2
For Use With (Application) COVID-19-immunoregulation
Synonym MC-207,110 dihydrochloride Phe-Arg-β-naphthylamide dihydrochloride
Solubility Information DMSO : 83.33 mg/mL (160.41 mM; Need ultrasonic) ∣H2O : 14.29 mg/mL (27.51 mM; Need ultrasonic)
SMILES O=C([C@H](CCCNC(N)=N)NC([C@H](CC1=CC=CC=C1)N)=O)NC2=CC(C=CC=C3)=C3C=C2.Cl.Cl
Molecular Weight (g/mol) 519.47
Formula Weight 519.47
Percent Purity 98.51%
Grade Research