Learn More
Thermo Scientific™ Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs
Product Code. 11902965 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Double-sided Comb
Prep Comb
Rapid Load Comb
Standard Comb
Wall Comb
Teeth Count:
1
12
16
2
20
24
25 (2X)
8
9 (1X)
Thickness (Metric) Teeth:
1 mm
1.0/1.5 mm
1.5 mm
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Teeth Count Thickness (Metric) Teeth unitSize
11902965 Double-sided Comb 16 1.0/1.5 mm Each
11952965 Double-sided Comb 12 1.0/1.5 mm Each
11912965 Double-sided Comb 20 1.0/1.5 mm Each
11922965 Double-sided Comb 24 1.0/1.5 mm Each
12474567 Standard Comb 8 1 mm Each
11983509 Standard Comb 8 1.5 mm Each
11962965 Prep Comb 2 1.5 mm Each
12404577 Rapid Load Comb 25 (2X) 1 mm Each
12414577 Rapid Load Comb 25 (2X) 1.5 mm Each
12424577 Rapid Load Comb 9 (1X) 1 mm Each
12434577 Rapid Load Comb 9 (1X) 1.5 mm Each
12444577 Wall Comb 1 1.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 11902965 Supplier Thermo Scientific™ Supplier No. B216
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs.
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with the EasyCast™ B2 and B3 Mini Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Ordering Information:
Cat. Nos. B2-RL-9C, -9D, -25C and -25D are 8 and 12-channel pipette format (1X) and (2X) combs
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl™ EasyCast B2 and B3 Mini Gel Electrophoresis System
Gel Size Mini
Product Line Owl, EasyCast
Type Double-sided Comb
Teeth Count 16
Thickness (Metric) Teeth 1.0/1.5 mm