Learn More
Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe
Product Code. 10654603
Change view
Click to view available options
For Use With (Equipment):
Orion DO Probes 080510MD, 081010MD, 081010
Orion DO Probes 083005MD, 083010MD, 083025MD, 083060MD, 086030MD, 083005A, 083010A, 083005D, 086020A, 081010F
Quantity:
1 Cap
3 Caps
Unit Size:
Each
Pack of 3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Quantity Unit Size
10654603 Orion DO Probes 083005MD, 083010MD, 083025MD, 083060MD, 086030MD, 083005A, 083010A, 083005D, 086020A, 081010F 1 Cap Each
11767757 Orion DO Probes 080510MD, 081010MD, 081010 3 Caps Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10654603 Supplier Thermo Scientific™ Supplier No. 080515
Fecha estimada de envoi: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Reduce probe servicing time and eliminate assembly problems using the Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe.
Reduce probe servicing time and eliminate assembly problems using Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe. Convenient screw-on membrane caps help ensure optimal probe performance.
Warranty: Out of box
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Orion DO Probes 083005MD, 083010MD, 083025MD, 083060MD, 086030MD, 083005A, 083010A, 083005D, 086020A, 081010F
Type Membrane Cap
Probe Type Polarographic DO
Quantity 1 Cap
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.