Learn More
S100 calcium binding protein A14 Antibody (S100A14/9077R), Alexa Fluor™ 594, R&D Systems™
Product Code. 18612090 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Product Code. Quantity Unit Size
18612090 0.1 mL 0.10mL
1 options
This item is not returnable. View return policy
Product Code. 18612090 Supplier Novus Biologicals Supplier No. NBP324305AF594
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody
S100 calcium binding protein A14 Monoclonal antibody specifically detects S100 calcium binding protein A14 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Antigen S100 calcium binding protein A14
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S100A14/9077R
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias protein S100-A14, S100 calcium binding protein A14, S100 calcium-binding protein A14, S100A15BCMP84breast cancer membrane protein 84, S114
Host Species Rabbit
Immunogen Recombinant full-length human S100 calcium binding protein A14 protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 57402
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Product Line Novus