Learn More
Nystatin, BLD Pharm™
Specifications
Specifications
| CAS | 1400-61-9 |
| Molecular Formula | C47H75NO17 |
| MDL Number | MFCD00036240 |
| Quantity | 25 g |
| SMILES | C[C@@H]1OC(C[C@H](O)C[C@H](O)C[C@H](O)CC[C@@H](O)[C@H](O)C[C@@](O)(O[C@H]2C[C@H](/C=C/C=C/C=C/C=C/CC/C=C/C=C/[C@@H]([C@@H](O)[C@H]1C)C)O[C@H]3[C@@H](O)[C@@H](N)[C@H](O)[C@@H](C)O3)C[C@H](O)[C@H]2C(O)=O)=O |
| Molecular Weight (g/mol) | 926.09 |
| Chemical Name or Material | Nystatin |