Learn More
NSC 663284, MedChemExpress
Product Code. 16495497
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495497 5 mg 5mg
16494216 100 mg 100mg
16492516 25 mg 25mg
16489456 50 mg 50mg
16482758 10 mg 10mg
16449995 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16495497 Supplier Medchem Express Supplier No. HY100034
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress NSC 663284 (DA-3003-1) is a potent, cell-permeable, and irreversible Cdc25 dual specificity phosphatase inhibitor, has an IC50 for Cdc25B2 of 0.21 μM. NSC 663284 exhibits mixed competitive kinetics against Cdc25A, Cdc25B(2), and Cdc25C with Ki values of 29, 95, and 89 nM, respectively. NSC 663284 inhibits NSD2 (IC50 of 170 nM) through a direct interaction with the catalytic SET domain (Kd of 370 nM).
NSC 663284 (DA-3003-1) is a potent, cell-permeable, and irreversible Cdc25 dual specificity phosphatase inhibitor, has an IC50 for Cdc25B2 of 0.21 μM. NSC 663284 exhibits mixed competitive kinetics against Cdc25A, Cdc25B(2), and Cdc25C with Ki values of 29, 95, and 89 nM, respectively. NSC 663284 inhibits NSD2 (IC50 of 170 nM) through a direct interaction with the catalytic SET domain (Kd of 370 nM).
Specifications
Chemical Name or Material NSC 663284
Physical Form Solid
CAS 383907-43-5
Color Red
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C15H16ClN3O3
For Use With (Application) Cancer-programmed cell death
Synonym DA-3003-1
Solubility Information DMSO : ≥ 150 mg/mL (466.19 mM)
SMILES O=C(C(Cl)=C1NCCN2CCOCC2)C3=C(N=CC=C3)C1=O
Molecular Weight (g/mol) 321.76
Formula Weight 321.76
Percent Purity 99.87%
Grade Research