Learn More
Thermo Scientific™ North2South™ Complete Biotin Random Prime Labeling and Detection Kit
Product Code. 10249423 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2164 mL Kit
Unit Size:
1 set
Product Code. Quantity unitSize
10249423 2164 mL Kit 1 set
1 options
This item is not returnable. View return policy
Product Code. 10249423 Supplier Thermo Scientific™ Supplier No. 17175
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Acquire consistent, highly sensitive Southern blot results with a novel ECL HRP substrate and optimized hybridization and blocking buffers.
Thermo Scientific™ North2South Chemiluminescent Detection Kit combines a novel ECL substrate for horseradish peroxidase (HRP) and optimized hybridization and blocking buffers to ensure consistent, high-sensitivity Southern blot results.
The Thermo Scientific North2South Hybridization and Detection Kit combines a novel enhanced luminol substrate for horseradish peroxidase (HRP) with optimized pre-made hybridization and blocking buffers to ensure consistent results and high sensitivity. Any nucleic acid blot can be hybridized with the appropriate biotinylated (biotin-labeled) oligonucleotide probe. After a brief membrane blocking step, the biotin label is detected with streptavidin-HRP and a 5-minute incubation with the chemiluminescent substrate. Results are easily recorded by brief exposure to X-ray film (1-15 minutes) or CCD camera.
Highlights:
  • Sensitivity equal to or greater than 32P
  • Amenable to many formats
  • High level of light output and consistent low background allows for very short exposure times using cooled CCD camera systems or film
  • Ready-to-use buffers and short process time makes switching from radioactive detection to chemiluminescent detection an easy transition, even for the novice user
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Sufficient For: 10 biotin-labeling reactions (each with 100 ng DNA) and 10 mini-gel blots
• North2South Chemiluminescent Hybridization and Detection Kit (Part No. 17097), 1 kit
• North2South Biotin Random Prime Labeling Kit (Part No. 17075), 1 kit
Detection Method Chromogenic
Format Kit
Product Line North2South
Product Type Biotin Labeling and Detection Kit
Quantity 2164 mL Kit