Learn More
Naphthalene-1,4-dicarboxylic acid, 98%, BLD Pharm™
Product Code. 31108697
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31108697 25 g
31108278 500 g
31103548 5 g
31099040 1 kg
31100357 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31108697 Supplier BLD Pharm Supplier No. BD1440725g
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 605-70-9
Molecular Formula C12H8O4
MDL Number MFCD00014312
Quantity 25 g
SMILES O=C(C1=C2C=CC=CC2=C(C(O)=O)C=C1)O
Molecular Weight (g/mol) 216.19
Percent Purity 98%
Chemical Name or Material Naphthalene-1,4-dicarboxylic acid