Learn More
N,N-Diethyl-2-acrylamide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2675-94-7 |
| Molecular Formula | C7H13NO |
| MDL Number | MFCD00078408 |
| Quantity | 25 g |
| SMILES | C=CC(N(CC)CC)=O |
| Molecular Weight (g/mol) | 127.18 |
| Percent Purity | 98% |
| Chemical Name or Material | N,N-Diethyl-2-acrylamide |