Learn More
N-Eicosane, 99%, BLD Pharm™
Product Code. 31033840
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
1kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31033840 500 g -
31030043 25 g -
31030204 1 kg 1kg
31025524 100 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31033840 Supplier BLD Pharm Supplier No. BD144090500g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 112-95-8
Molecular Formula C20H42
MDL Number MFCD00009344
Quantity 500 g
SMILES CCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 282.55
Percent Purity 99%
Chemical Name or Material N-Eicosane