missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-oxo-L-proline tert-butyl ester, 97%

Product Code. 11367159
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367159 5 g 5g
15423677 250 mg 250mg
11357159 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11367159 Supplier Thermo Scientific Alfa Aesar Supplier No. H27375.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 166410-05-5 | C14H23NO5 | 285.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166410-05-5
Molecular Formula C14H23NO5
Molecular Weight (g/mol) 285.34
MDL Number MFCD06797013
InChI Key MPNWQUWKRDADHK-JTQLQIEISA-N
Synonym n-boc-4-oxo-l-proline tert-butyl ester, s-di-tert-butyl 4-oxopyrrolidine-1,2-dicarboxylate, boc-4-oxo-l-proline tert-butyl ester, 1,2-di-tert-butyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, boc-4-oxo-l-prolinetert-butylester, di-tert-butyl s-4-oxopyrrolidine-1,2-dicarboxylate, di-tert-butyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, 1-tert-butoxycarbonyl-4-oxo-l-proline tert-butyl ester, 1-tert-butyloxycarbonyl-4-oxo-l-proline tert-butyl ester, s-4-oxopyrrolidine-1,2-dicarboxylic acid di-tert-butyl ester
PubChem CID 11346670
IUPAC Name ditert-butyl (2S)-4-oxopyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)C1CC(=O)CN1C(=O)OC(C)(C)C

Specifications

Melting Point 64°C to 67°C
Quantity 5 g
Formula Weight 285.35
Percent Purity 97%
Chemical Name or Material N-Boc-4-oxo-L-proline tert-butyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.