missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3,4-dichloro-D-phenylalanine, 95%, Thermo Scientific™

Product Code. 15498587
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15498587 250 mg 250mg
15478587 1 g 1g
15488587 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 15498587

Brand: Thermo Scientific Alfa Aesar H52070.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114873-13-1
Molecular Formula C14H16Cl2NO4
Molecular Weight (g/mol) 333.19
MDL Number MFCD00151887
InChI Key UGZIQCCPEDCGGN-LLVKDONJSA-M
Synonym boc-3,4-dichloro-d-phenylalanine, boc-d-phe 3,4-cl2-oh, r-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, boc-d-3,4-dichlorophe, boc-d-3,4-dichlorophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, boc-d-phe 3,4-dicl-oh, boc-3,4-dichloro-d-phe-oh, n-boc-d-3,4-dichlorophenylalanine, d-phenylalanine, 3,4-dichloro-n-1,1-dimethylethoxy carbonyl
PubChem CID 7015811
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C(Cl)=C1)C([O-])=O

Specifications

Quantity 250 mg
Beilstein 8428163
IUPAC Name (2R)-3-(3,4-dichlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Formula Weight 334.2
Percent Purity 95%
Chemical Name or Material N-Boc-3,4-dichloro-D-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.