Learn More
N-(2-Butyl-3-(4-(3-(dibutylamino)propoxy)benzoyl)benzofuran-5-yl)methanesulfonamide hydrochloride, 95%, BLD Pharm™
Product Code. 31091039
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31091039 1 g
31078310 10 g
31078692 5 g
31079995 250 mg
31084183 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31091039 Supplier BLD Pharm Supplier No. BD1269541g
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 141625-93-6
Molecular Formula C31H45ClN2O5S
MDL Number MFCD00914940
Quantity 1 g
SMILES CS(=O)(NC1=CC=C(OC(CCCC)=C2C(C3=CC=C(OCCCN(CCCC)CCCC)C=C3)=O)C2=C1)=O.[H]Cl
Molecular Weight (g/mol) 593.22
Percent Purity 95%
Chemical Name or Material N-(2-Butyl-3-(4-(3-(dibutylamino)propoxy)benzoyl)benzofuran-5-yl)methanesulfonamide hydrochloride