missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ LB-Agar, Powdered

Product Code. 11303269 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
454 g
1000 g
5000 g
10 x 500 mL
10 x 1000 mL
Form:
Powder
Content And Storage:
15°C to 30°C
Room Temperature
Unit Size:
10 L
1kg
454g
5 L
5kg
Product Code. Quantity Form Content And Storage unitSize
11303269 5000 g Powder 15°C to 30°C 5kg
11383259 1000 g Powder 15°C to 30°C 1kg
11373269 10 x 1000 mL Powder Room Temperature 10 L
11363269 10 x 500 mL Powder Room Temperature 5 L
11353259 454 g Powder 15°C to 30°C 454g
12968326 1000 g Powder 15°C to 30°C 1kg
6 options
This item is not returnable. View return policy

Product Code. 11303269

Brand: MP Biomedicals™ 113002241

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Offered in a variety of formats for convenience and ease of use

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™ FDA
Content And Storage 15°C to 30°C
Form Powder
Format Capsule
Grade Molecular biology
Product Type Agar
Formulation Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L
For Use With (Application) For growth of bacteria
Quantity 5000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.