Learn More
Mouse kidney tissue lysate (14 days), 100 ug
Product Code. 32157063 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
100µg
Product Code. Unit Size
32157063 100µg
1 options
This item is not returnable. View return policy
Product Code. 32157063 Supplier Abcam Supplier No. ab7262100ug
 Restricted Item
Fecha estimada de envoi: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Mouse kidney cell/tissue lysate.
  • Species: Mouse
  • Tissue: kidney
  • Form: Liquid
Specifications
For Use With (Application) Western Blot