Learn More
Minoxidil, Thermo Scientific™
Minoxidil, CAS # 38304-91-5, is a comopund with vasodilating effects that works by opening adenosine triphosphate (ATP)-sensitive potassium channels.
37.00€ - 251.00€
CAS 38304-91-5
Fórmula molecular C9H15N5O
Peso molecular (g/mol) 209.253
Número MDL MFCD00063409
Clave InChI ZIMGGGWCDYVHOY-UHFFFAOYSA-N
Sinónimo minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox
PubChem CID 4201
Nombre IUPAC 3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine
SMILES C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O
Ver más especificaciones
Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15444319
SDS Ver documentos
Thermo Scientific Alfa Aesar
J61803.03
1g
37.00€
1 g
Ver productos alternativos
Ahorre hasta 
»
15454319
SDS Ver documentos
Thermo Scientific Alfa Aesar
J61803.06
5g
ONLINE EXCLUSIVE PRICE
121.00€
108.65€ / 5 g
Ahorro 12.35€
10% Descuento
Ver productos alternativos
Ahorre hasta 
»
15464319
SDS Ver documentos
Thermo Scientific Alfa Aesar
J61803.14
25g
251.00€
25 g
Ver productos alternativos
Ahorre hasta 
»
Descripción
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Minoxidil is a piperidine that acts as a vasodilator when converted into its active metabolite, minoxidil sulphate
• Minoxidil opens adenosine triphosphate (ATP)-sensitive potassium channels in the plasma membrane and relaxes smooth muscle in arteriole walls. Relaxation of smooth muscle within the arterioles leads to decreased resistance and lower blood pressure
Application
• Minoxidil metabolism is a common focus of its use in research because of the number of enzymes involved in its sulfation and clearance
• Minoxidil can be used for in vivo or in vitro physiology research
Especificaciones
38304-91-5
209.253
ZIMGGGWCDYVHOY-UHFFFAOYSA-N
4201
C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O
C9H15N5O
MFCD00063409
minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox
3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine
Especificaciones
Minoxidil
C9H15N5O
14,6203
Soluble in water (2.2mg/ml),100% ethanol (29mg/ml),propylene glycol,acetone,DMSO (6.5mg/ml),and methanol.
C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O
209.253
209.25
38304-91-5
MFCD00063409
minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox
ZIMGGGWCDYVHOY-UHFFFAOYSA-N
3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine
4201
Videos
Seguridad y manipulación
Seguridad y manipulación
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 253-874-2
missing translation for 'rtecsNumber' : UV8200000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures