Learn More
MFF Antibody, Biotin, R&D Systems™
Product Code. 18603787 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Product Code. Quantity Unit Size
18603787 0.1 mL 0.10mL
1 options
This item is not returnable. View return policy
Product Code. 18603787 Supplier Novus Biologicals Supplier No. NBP318442B
 Restricted Item
Fecha estimada de envoi: 01-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
MFF Polyclonal specifically detects MFF in Human samples. It is validated for Immunoprecipitation∣Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Antigen MFF
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Formulation PBS
Gene Alias mitochondrial fission factor
Host Species Rabbit
Immunogen Recognizes region between residues Between Between 50 and 100..
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 56947
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus