Learn More
Methyl Phenylpropiolate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 4891-38-7 |
| Molecular Formula | C10H8O2 |
| MDL Number | MFCD00041685 |
| Quantity | 25 g |
| SMILES | O=C(OC)C#CC1=CC=CC=C1 |
| Molecular Weight (g/mol) | 160.17 |
| Percent Purity | 98% |
| Chemical Name or Material | Methyl Phenylpropiolate |