Learn More
Methyl 3-methoxy-2-naphthoate, 96%, BLD Pharm™
Product Code. 31091385
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31091385 10 g
31084625 25 g
31089113 250 mg
31082042 5 g
31086027 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31091385 Supplier BLD Pharm Supplier No. BD13870810g
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 13041-60-6
Molecular Formula C13H12O3
MDL Number MFCD00454253
Quantity 10 g
SMILES O=C(OC)C1=C(OC)C=C2C=CC=CC2=C1
Molecular Weight (g/mol) 216.23
Percent Purity 96%
Chemical Name or Material Methyl 3-methoxy-2-naphthoate