Learn More
Methyl 2-(triphenylphosphoranylidene)acetate, 98%, BLD Pharm™
Product Code. 30937942
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30937942 10 g 10g
30932064 100 g 100g
30931228 25 g 25g
30936250 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30937942 Supplier BLD Pharm Supplier No. BD133410g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 2605-67-6
Molecular Formula C21H19O2P
MDL Number MFCD00008455
Quantity 10 g
SMILES C3=C([P](C1=CC=CC=C1)(C2=CC=CC=C2)=CC(OC)=O)C=CC=C3
Molecular Weight (g/mol) 334.35
Percent Purity 98%
Chemical Name or Material Methyl 2-(triphenylphosphoranylidene)acetate