Learn More
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Product Code. 10240535
Change view
Click to view available options
Size:
8 mm
11 mm
13 mm
20 mm
28 mm
30 mm
32 mm
Type:
Manual Crimper
Manual Crimper (Flip-off)
Manual Decapping Plier
Manual Decrimper
Unit Size:
Each
Product Code. Size Type unitSize
10240535 13 mm Manual Decrimper Each
15280389 8 mm Manual Crimper Each
14860742 11 mm Manual Crimper Each
13488988 20 mm Manual Crimper Each
10147664 8 mm Manual Crimper Each
10429454 11 mm Manual Crimper Each
10566765 13 mm Manual Crimper Each
10231585 13 mm Manual Crimper (Flip-off) Each
12400268 20 mm Manual Crimper Each
10372525 20 mm Manual Crimper (Flip-off) Each
10250535 28 mm Manual Crimper Each
10773667 30 mm Manual Crimper Each
10229964 32 mm Manual Crimper Each
11528470 11 mm Manual Crimper Each
11538470 20 mm Manual Crimper Each
15290389 8 mm - Each
15188784 20 mm - Each
10259614 8 mm Manual Decrimper Each
10488084 11 mm Manual Decrimper Each
10279364 20 mm Manual Decrimper Each
12852344 28 mm Manual Decrimper Each
10301535 30 mm Manual Decrimper Each
11528240 32 mm Manual Decrimper Each
11564815 20 mm Manual Crimper Each
10530303 11 mm Manual Decrimper Each
10143362 20 mm Manual Decrimper Each
12054907 11 mm Manual Decapping Plier Each
10299454 13 mm Manual Decapping Plier Each
10066164 20 mm Manual Decapping Plier Each
10200205 30 mm Manual Decapping Plier Each
30 options
This item is not returnable. View return policy
Product Code. 10240535
Brand: Thermo Scientific™ C4013102
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Manual Crimpers
Lightweight, precision crimpers attach seals to chromatography or serum vials. Available for standard aluminum or flip-off seals.
  • Factory set to properly crimp standard-thickness seals
  • Adjustable handle setting screw allows proper crimps with septa of varying thickness
Manual Decripmers
Forged pliers feature a fitted gripping surface designed to allow removal of the listed sizes of seals.
  • Simply grip pliers around top of seal, squeeze and pry upward to remove
Manual Decrimping Pliers
Place decrimping head squarely on seal and squeeze.
  • Minimizes damage to vial rim with chromatography or serum vials
  • Removes standard aluminum seals and many steel seals
TRUSTED_SUSTAINABILITY
Specifications
Description Manual Decrimper
No. per Pack 1 Ea.
Type Manual Decrimper
For Use With (Equipment) 13 mm Standard Seal, National Vial, Chromacol Vial
Size 13 mm
For Use With (Application) Industrial Chromatography
Product Line National
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.