Learn More
Lymphatic Endothelial Cells Antibody (LA102), mFluor Violet 610 SE, R&D Systems™
Product Code. 18609819 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Product Code. Quantity Unit Size
18609819 0.1 mL 0.10mL
1 options
This item is not returnable. View return policy
Product Code. 18609819 Supplier Novus Biologicals Supplier No. NB11096416MFV610
 Restricted Item
Fecha estimada de envoi: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Novus product, part of R&D Systems. Rat Monoclonal Antibody
Lymphatic Endothelial Cells Monoclonal specifically detects Lymphatic Endothelial Cells in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Antigen Lymphatic Endothelial Cells
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone LA102
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias LEC26
Host Species Rat
Immunogen Collagenase- and neuraminidase-treated mouse benign lymphangiomas
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line Novus