missing translation for 'onlineSavingsMsg'
Learn More

LPO Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16856723 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16856723 150 μL 150µL
16866723 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16856723

Brand: Proteintech 103761AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes an oxidoreductase secreted from salivary, mammary, and other mucosal glands that functions as a natural antibacterial agent. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen LPO
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene LPO
Gene Accession No. P22079
Gene Alias lactoperoxidase, LPO, Salivary peroxidase, SAPX, SPO
Gene Symbols LPO
Host Species Rabbit
Immunogen peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4025, 76113
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.