Learn More
Lipoamide, MedChemExpress
Product Code. 16488636
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
16488636 100 mg 100mg
1 options
This item is not returnable. View return policy
Product Code. 16488636 Supplier Medchem Express Supplier No. HYB1142
 Restricted Item
Fecha estimada de envoi: 23-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Lipoamide is a coenzyme, which transfer acetyl and hydrogen in Pyruvate deacylation oxidation Process, used for pharmaceuticals.
Lipoamide is a coenzyme, which transfer acetyl and hydrogen in Pyruvate deacylation oxidation Process, used for pharmaceuticals.
Specifications
Chemical Name or Material Lipoamide
CAS 940-69-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C8H15NOS2
Quantity 100 mg
Synonym (±)-α-Lipoamide DL-Lipoamide DL-6,8-Thioctamide
Solubility Information DMSO : ≥ 50 mg/mL (243.50 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N)CCCCC1SSCC1
Molecular Weight (g/mol) 205.34
Formula Weight 205.34
Percent Purity 96.04%
Grade Research
Physical Form Solid