missing translation for 'onlineSavingsMsg'
Learn More

L-Glycine 7-amido-4-methylcoumarin, Thermo Scientific Chemicals

Product Code. 15426989
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
Product Code. Quantity unitSize
15426989 10 mg 10mg
15436989 25 mg 25mg
2 options
This item is not returnable. View return policy

Product Code. 15426989

Brand: Thermo Scientific Alfa Aesar J65592.MA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12H12N2O3
MDL Number MFCD00077122
Synonym L-Gly-AMC; L-G-AMC

Specifications

Color White
Quantity 10 mg
Sensitivity Light sensitive
Solubility Information Soluble in DMSO or water
Formula Weight 232.23
Physical Form Solid
Chemical Name or Material L-Glycine 7-amido-4-methylcoumarin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.