Learn More
Lonza™ KGM™ Gold Keratinocyte Growth Medium SingleQuots™ Supplements and Growth Factors
Product Code. 11615090
Change view
Click to view available options
Quantity:
0.5 to 2.0 mL ViaLs
Unit Size:
Each
Product Code. Quantity Unit Size
11615090 0.5 to 2.0 mL ViaLs Each
1 options
This item is not returnable. View return policy
Product Code. 11615090 Supplier Lonza™ Supplier No. 192152
  • ONLINE EXCLUSIVE PRICE
    291.00€
    227.62€ / paquete de 1
    Ahorro 63.38€
    21% Descuento
View all promotions
 Restricted Item
Fecha estimada de envoi: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Separately sold supplement and growth factor set for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures.
  • Supplement and growth factor set sold separately from basal media.
  • Can be used with 1 bottle of 192060 (basal media).
  • Designed to support both clonal growth and high density keratinocyte proliferation.
  • Supports superior growth and morphology of keratinocytes.
  • 1 x KBM™Gold™ Basal Medium (00192151), 500 mL
  • 1 x KGM™Gold™SingleQuots™Supplement pack (00192152) containing:
    1 x Natural Cap Vial with Hydrocortisone, 0.50 mL
    1 x Natural Cap Vial with Transferrin, 0.50 mL
    1 x Amber Vial with Epinephrine, 0.25 mL
    1 x Red Cap Vial with GA-1000, 0.50 mL
    1 x Orange Cap Vial with BPE, 2.00 mL
    1 x Green Cap Vial with hEGF, 0.50 mL
    1 x Lilac Cap Vial with Insulin, 0.50 mL
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) High and low density NHEK-Normal Human Epidermal Keratinocyte cultures
Quantity 0.5 to 2.0 mL ViaLs