Learn More
KD Scientific Splitters
Directs airflow into and out of vials to facilitate solvent evaporation
Descripción
- For use with Centrfian micro centrifuge vial rotor/micro test tube rotor/vial rotor/scintillating vial rotor/10 HPLC vial rotor
Especificaciones
| Splitter | |
| Polyethylene |