Learn More
Isobutyltriphenylphosphonium bromide, 98%, BLD Pharm™
Product Code. 30948932
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
Unit Size:
100g
1g
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948932 100 g 100g
30931130 5 g 5g
30935470 25 g 25g
30930840 1 g 1g
30939374 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30948932 Supplier BLD Pharm Supplier No. BD1491100g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 22884-29-3
Molecular Formula C22H24BrP
MDL Number MFCD00031593
Quantity 100 g
SMILES CC(C)C[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
Molecular Weight (g/mol) 399.30
Percent Purity 98%
Chemical Name or Material Isobutyltriphenylphosphonium bromide